Learn More
53BP1 Antibody - BSA Free, Novus Biologicals™
Product Code. 18799923 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mg
25µg
Quantity:
0.025 mL
Product Code. unitSize Quantity
18799923 0.1mg -
18735403 25µg 0.025 mL
2 options
Product Code. 18799923
Brand: Novus Biologicals NB100304
Rabbit Polyclonal Antibody
53BP1 Polyclonal specifically detects 53BP1 in Human, Mouse, Rat, Bat, Bovine, Canine, Fish, Goat, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, In-situ Hybridization, Flow (Intracellular), Knockout Validated, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Gene Alias 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1
Gene Symbols TP53BP1