missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK1 Antibody (SR2210), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21577-50ul
This item is not returnable.
View return policy
Description
IRAK1 Monoclonal antibody specifically detects IRAK1 in Human samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation
Specifications
| IRAK1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| SR2210 | |
| Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 1, IRAK-1, IRAKPelle homolog, pelle | |
| A synthesized peptide derived from human IRAK1 (Uniprot #: P51617) | |
| 50 μg | |
| Protein Kinase | |
| 3654 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction