missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK1 Antibody (SR2210), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | IRAK1 |
|---|---|
| Clone | SR2210 |
| Dilution | Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18601535
|
Novus Biologicals
NBP3-21577-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18667954
|
Novus Biologicals
NBP3-21577-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IRAK1 Monoclonal antibody specifically detects IRAK1 in Human samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
| IRAK1 | |
| Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 1, IRAK-1, IRAKPelle homolog, pelle | |
| A synthesized peptide derived from human IRAK1 (Uniprot #: P51617) | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| SR2210 | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 3654 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title