missing translation for 'onlineSavingsMsg'
Learn More

4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}phenol, TRC

Product Code. 30005206
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005206 50 mg 50mg
30134802 500 mg 500mg
30136645 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005206 Supplier Toronto Research Chemicals Supplier No. TRCT90128350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 69045-85-8
Molecular Formula C12H8NO2F3
Molecular Weight (g/mol) 255.19
Synonym 4-[[5-(Trifluoromethyl)-2-pyridinyl]oxy]phenol2-(4-Hydroxyphenoxy)-5-(trifluoromethyl)pyridine4-(5-Trifluoromethyl-2-pyridyloxy)phenol4-(5-Trifluoromethylpyridin-2-yloxy)phenol
IUPAC Name 4-[5-(trifluoromethyl)pyridin-2-yl]oxyphenol
SMILES Oc1ccc(Oc2ccc(cn2)C(F)(F)F)cc1

Tekniske data

Quantity 50 mg
InChI Formula InChI=1S/C12H8F3NO2/c13-12(14,15)8-1-6-11(16-7-8)18-10-4-2-9(17)3-5-10/h1-7,17H
Formula Weight 255.0507
Chemical Name or Material 4-([5-(Trifluoromethyl)pyridin-2-yl]oxy)phenol
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.