CAS RN 69045-85-8
CAS RN 69045-85-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}phenol, TRC
CAS: 69045-85-8 Molecular Formula: C12H8NO2F3 Molecular Weight (g/mol): 255.19 Synonym: 4-[[5-(Trifluoromethyl)-2-pyridinyl]oxy]phenol2-(4-Hydroxyphenoxy)-5-(trifluoromethyl)pyridine4-(5-Trifluoromethyl-2-pyridyloxy)phenol4-(5-Trifluoromethylpyridin-2-yloxy)phenol IUPAC Name: 4-[5-(trifluoromethyl)pyridin-2-yl]oxyphenol SMILES: Oc1ccc(Oc2ccc(cn2)C(F)(F)F)cc1