missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethyl)-2-furaldehyde, ≥97%, Thermo Scientific™

Product Code. 10759605
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10759605 250 mg Amber Glass Bottle 250mg
10438423 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10759605 Supplier Thermo Scientific Maybridge Supplier No. CC49804CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 17515-80-9
Molecular Formula C6H3F3O2
Molecular Weight (g/mol) 164.083
MDL Number MFCD00525585
InChI Key KLVCIHFXPHKSPD-UHFFFAOYSA-N
Synonym 5-trifluoromethyl furan-2-carbaldehyde, 5-trifluoromethyl-2-furaldehyde, 2-furancarboxaldehyde, 5-trifluoromethyl, 2-furancarboxaldehyde,5-trifluoromethyl, 5-trifluoromethyl furfural, 2-formyl-5-trifluoromethyl furan, 5-trifluoromethyl-furan-2-carbaldehyde, 5-trifluoromethyl-2-furancarboxaldehyde, 5-trifluoromethyl furan-2-carboxaldehyde
PubChem CID 7537665
IUPAC Name 5-(trifluoromethyl)furan-2-carbaldehyde
SMILES C1=C(OC(=C1)C(F)(F)F)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 164.08
Percent Purity 97%
Chemical Name or Material 5-(Trifluoromethyl)-2-furaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.