accessibility menu, dialog, popup

UserName

OFFFO

CAS RN 17515-80-9

IUPAC Name: 5-(trifluoromethyl)furan-2-carbaldehyde
Synonyms: 5-(trifluoromethyl)furan-2-carbaldehyde
Molecular Weight (g/mol): 164.08
Molecular Formula: C6H3F3O2
InChi Key: KLVCIHFXPHKSPD-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(O1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results