missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornen-2-yl acetate, mixture of endo and exo, 98%

Product Code. 15472966
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15472966

Brand: Thermo Scientific Alfa Aesar H56907.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6143-29-9 | C9H12O2 | 152.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6143-29-9
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00167565
InChI Key DRWRVXAXXGJZIO-UHFFFAOYSA-N
Synonym 5-norbornen-2-yl acetate, 5-norbornene-2-yl acetate, 2-acetoxy-5-norbornene, bicyclo 2.2.1 hept-5-en-2-yl acetate, bicyclo 2.2.1 hept-5-en-2-ol, acetate, acetic acid bicyclo 2.2.1 hept-5-en-2-yl ester, 5-norbornen-2-yl acetate, mixture of endo and exo, 5-norbonen-2-yl acetate, 5-norbornen-2-ol, acetate
PubChem CID 95688
IUPAC Name 5-bicyclo[2.2.1]hept-2-enyl acetate
SMILES CC(=O)OC1CC2CC1C=C2

Specifications

Density 1.044
Boiling Point 73°C to 76°C (14 mmHg)
Flash Point 62°C (144°F)
Refractive Index 1.47
Quantity 100 g
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 5-Norbornen-2-yl acetate, mixture of endo and exo

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.