accessibility menu, dialog, popup

UserName

CAS RN 6143-29-9

H3COO

CAS RN 6143-29-9

IUPAC Name: bicyclo[2.2.1]hept-5-en-2-yl acetate
Synonyms: bicyclo[2.2.1]hept-5-en-2-yl acetate
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: DRWRVXAXXGJZIO-UHFFFAOYNA-N
SMILES: CC(=O)OC1CC2CC1C=C2
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results