missing translation for 'onlineSavingsMsg'
Learn More

5-Nitropyridine-2-carboxylic acid, 97+%

Product Code. 15491739
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15491739 5 g 5g
15481739 1 g 1g
15401749 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491739 Supplier Thermo Scientific Alfa Aesar Supplier No. H64499.06
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 30651-24-2 | C6H4N2O4 | 168.11 g/mol

It plays an important role in synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It plays an important role in synthetic intermediate.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30651-24-2
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.11
MDL Number MFCD04114182
InChI Key QKYRCTVBMNXTBT-UHFFFAOYSA-N
Synonym 5-nitropicolinic acid, 5-nitropyridine-2-carboxylic acid, 5-nitro-2-pyridinecarboxylic acid, 5-nitro-2-picolinic acid, 2-pyridinecarboxylic acid, 5-nitro, picolinic acid, 5-nitro, 3-nitro-6-pyridine carboxylic acid, 2-carboxy-5-nitropyridine, 5-nitropyridine-2-carboxylicacid
PubChem CID 520488
IUPAC Name 5-nitropyridine-2-carboxylic acid
SMILES OC(=O)C1=NC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 208°C to 212°C
Quantity 5 g
Beilstein 139209
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 168.11
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 5-Nitropyridine-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.