CAS RN 30651-24-2
IUPAC Name:
5-nitropyridine-2-carboxylic acid
Synonyms:
5-nitropyridine-2-carboxylic acid
Molecular Weight (g/mol):
168.11
Molecular Formula:
C6H4N2O4
InChi Key:
QKYRCTVBMNXTBT-UHFFFAOYSA-N
SMILES:
OC(=O)C1=NC=C(C=C1)[N+]([O-])=O
1
–
1
of
1
results
1
5-Nitropyridine-2-carboxylic acid, 97+%
CAS: 30651-24-2 Molecular Formula: C6H4N2O4 Molecular Weight (g/mol): 168.11 MDL Number: MFCD04114182 InChI Key: QKYRCTVBMNXTBT-UHFFFAOYSA-N Synonym: 5-nitropicolinic acid,5-nitropyridine-2-carboxylic acid,5-nitro-2-pyridinecarboxylic acid,5-nitro-2-picolinic acid,2-pyridinecarboxylic acid, 5-nitro,picolinic acid, 5-nitro,3-nitro-6-pyridine carboxylic acid,2-carboxy-5-nitropyridine,5-nitropyridine-2-carboxylicacid PubChem CID: 520488 IUPAC Name: 5-nitropyridine-2-carboxylic acid SMILES: OC(=O)C1=NC=C(C=C1)[N+]([O-])=O