missing translation for 'onlineSavingsMsg'
Learn More

5-(N-Ethyl-N-2-hydroxyethylamino)-2-penthlamine, TRC

Product Code. 30132683
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.50g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132683 2.5 g 2.50g
30010578 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132683 Supplier Toronto Research Chemicals Supplier No. TRCE6788552.5G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69559-11-1
Molecular Formula C11 H26 N2 O
Molecular Weight (g/mol) 202.337
Synonym 2-[(4-Aminopentyl)ethylamino]-ethanol, (±)-2-[(4-Aminopentyl)ethylamino]ethanol, 5-[N-Ethyl-N-(2-hydroxyethyl)amino]-2-aminopentane, N-Ethyl-N-(2-hydroxyethyl)-4-aminopentylamine
IUPAC Name 2-[6-aminoheptyl(ethyl)amino]ethanol
SMILES CCN(CCO)CCCCCC(C)N

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C11H26N2O/c1-3-13(9-10-14)8-6-4-5-7-11(2)12/h11,14H,3-10,12H2,1-2H3
Formula Weight 202.2045
Chemical Name or Material 5-(N-Ethyl-N-2-hydroxyethylamino)-2-penthlamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.