CAS RN 69559-11-1
CAS RN 69559-11-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-(N-Ethyl-N-2-hydroxyethylamino)-2-penthlamine, TRC
CAS: 69559-11-1 Molecular Formula: C11 H26 N2 O Molecular Weight (g/mol): 202.337 Synonym: 2-[(4-Aminopentyl)ethylamino]-ethanol,(±)-2-[(4-Aminopentyl)ethylamino]ethanol,5-[N-Ethyl-N-(2-hydroxyethyl)amino]-2-aminopentane,N-Ethyl-N-(2-hydroxyethyl)-4-aminopentylamine IUPAC Name: 2-[6-aminoheptyl(ethyl)amino]ethanol SMILES: CCN(CCO)CCCCCC(C)N