missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-MethylCytosine Antibody, Novus Biologicals™
Sheep Polyclonal Antibody has been used in 3 publications
£175.00 - £305.00
Specifications
| Antigen | 5-MethylCytosine |
|---|---|
| Concentration | 3.0 mg/mL |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18268104
|
Novus Biologicals
NB100-744-0.025MG |
0.025 mg |
£175.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18768953
|
Novus Biologicals
NB100-744 |
0.125 mg |
£305.00
125µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-MethylCytosine Polyclonal specifically detects 5-MethylCytosine in Human, Rat, All Species samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| 5-MethylCytosine | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.5) with No Preservative | |
| Chemical Small Molecule conjugated to KLH. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 3.0 mg/mL | |
| Unconjugated | |
| Sheep | |
| All species, Rat | |
| 5-mC, 5-Methyl Cytosine, Cytosine (5-Methyl) | |
| IgG | |
| Protein G purified | |
| This is specific for 5-Methyl Cytosine. It was tested using immunoprecipitation against 5-Methyl Cytosine and bromodeoxyuridine (BrdU) or a control (no antigen). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title