missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-MethylCytosine Antibody, Novus Biologicals™
Sheep Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-744
This item is not returnable.
View return policy
Description
5-MethylCytosine Polyclonal specifically detects 5-MethylCytosine in Human, Rat, All Species samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| 5-MethylCytosine | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.5) with No Preservative | |
| Sheep | |
| Protein G purified | |
| RUO | |
| This is specific for 5-Methyl Cytosine. It was tested using immunoprecipitation against 5-Methyl Cytosine and bromodeoxyuridine (BrdU) or a control (no antigen). | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 3.0 mg/mL | |
| Western Blot, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 25 to 100 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL | |
| 5-mC, 5-Methyl Cytosine, Cytosine (5-Methyl) | |
| Chemical Small Molecule conjugated to KLH. | |
| 0.125 mg | |
| Primary | |
| Rat, All species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction