missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRDT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80294
This item is not returnable.
View return policy
Description
BRDT Polyclonal specifically detects BRDT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| BRDT | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| bromodomain testis-specific protein, bromodomain, testis-specific, Cancer/testis antigen 9, CT9BRD6, RING3-like protein | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| NP_001717 | |
| BRDT | |
| Synthetic peptide directed towards the N terminal of human BRDT. Peptide sequence MSLPSRQTAIIVNPPPPEYINTKKNGRLTNQLQYLQKVVLKDLWKHSFSW. | |
| 100 μL | |
| Protein Kinase | |
| 676 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction