missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-pyridinol, 97%, Thermo Scientific™

Product Code. 10167482
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10167482 1 g 1g
10604083 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10167482 Supplier Thermo Scientific Maybridge Supplier No. AC35768DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91914-06-6
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD03092888
InChI Key SOHMZGMHXUQHGE-UHFFFAOYSA-N
Synonym 2-hydroxy-5-methylpyridine, 5-methylpyridin-2-ol, 2-hydroxy-5-picoline, 5-methyl-2-pyridinol, 5-methylpyridin-2 1h-one, 5-methyl-2-pyridone, 5-methyl-2 1h-pyridinone, 2 1h-pyridinone, 5-methyl, 2-pyridinol, 5-methyl, 6-hydroxy-3-picoline
PubChem CID 70482
IUPAC Name 5-methyl-1H-pyridin-2-one
SMILES CC1=CNC(=O)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 180°C to 185°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 109.13
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-2-pyridinol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.