accessibility menu, dialog, popup

UserName

CH3NHO

CAS RN 91914-06-6

IUPAC Name: 5-methyl-1,2-dihydropyridin-2-one
Synonyms: 5-methyl-1H-pyridin-2-one
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: SOHMZGMHXUQHGE-UHFFFAOYSA-N
SMILES: CC1=CNC(=O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)