missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Lipoxygenase Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£186.00 - £400.00
Specifications
| Antigen | 5-Lipoxygenase |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Simple Western 50 ug/ml, Immunocytochemistry/Immunofluorescence 1:500 - 1:2000, KnockDown Validated |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18189809
|
Novus Biologicals
NB110-58748SS |
0.025 mL |
£186.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18229920
|
Novus Biologicals
NB110-58748 |
0.1 mL |
£400.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-Lipoxygenase Polyclonal specifically detects 5-Lipoxygenase in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
| 5-Lipoxygenase | |
| Western Blot 1:500-1:1000, Simple Western 50 ug/ml, Immunocytochemistry/Immunofluorescence 1:500 - 1:2000, KnockDown Validated | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| 5-lipoxygenase, 5-LO, 5-LOX, 5LPG, arachidonate 5-lipoxygenase, arachidonic 5-lipoxygenase alpha-10 isoform, arachidonic 5-lipoxygenase delta-10-13 isoform, arachidonic 5-lipoxygenase delta-13 isoform, arachidonic 5-lipoxygenase delta-p10 isoform, arachidonic acid 5-lipoxygenase, EC 1.13.11, EC 1.13.11.34, leukotriene A4 synthase, LOG5, MGC163204 | |
| ALOX5 | |
| IgG | |
| Affinity Purified | |
| 80 kDa |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| P09917 | |
| 240 | |
| A synthetic peptide made to a region within residues 100-200 of human 5-Lipoxygenase. [Swiss-Prot# P09917] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title