missing translation for 'onlineSavingsMsg'
Learn More

5-Lipoxygenase Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications

Brand:  Novus Biologicals NB110-58748SS

 View more versions of this product

Product Code. 18189809

  • £186.00 / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

5-Lipoxygenase Polyclonal specifically detects 5-Lipoxygenase in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5-Lipoxygenase
Polyclonal
Unconjugated
PBS and 30% Glycerol with 0.1% Sodium Azide
5-lipoxygenase, 5-LO, 5-LOX, 5LPG, arachidonate 5-lipoxygenase, arachidonic 5-lipoxygenase alpha-10 isoform, arachidonic 5-lipoxygenase delta-10-13 isoform, arachidonic 5-lipoxygenase delta-13 isoform, arachidonic 5-lipoxygenase delta-p10 isoform, arachidonic acid 5-lipoxygenase, EC 1.13.11, EC 1.13.11.34, leukotriene A4 synthase, LOG5, MGC163204
Rabbit
80 kDa
0.025 mL
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
240
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown
1 mg/mL
Western Blot 1:500-1:1000, Simple Western 50 ug/ml, Immunocytochemistry/Immunofluorescence 1:500 - 1:2000, KnockDown Validated
P09917
ALOX5
A synthetic peptide made to a region within residues 100-200 of human 5-Lipoxygenase. [Swiss-Prot# P09917]
Affinity Purified
RUO
Primary
Human, Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only