missing translation for 'onlineSavingsMsg'
Learn More

5-Keto-D-gluconic Acid, TRC

Product Code. 30397153
Change view
Click to view available options
Quantity:
25 mg
50 mg
200 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397153 25 mg
30396857 50 mg
30406105 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397153 Supplier Toronto Research Chemicals Supplier No. TRCK19131525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5287-64-9
Molecular Formula C6H10O7
Molecular Weight (g/mol) 194.14
Synonym D-xylo-5-Hexulosonic acid, 5-Oxo-D-gluconic acid, 5-Keto-D-gluconic acid, 5-keto-D-Gluconic acid
IUPAC Name (2R,3S,4S)-2,3,4,6-tetrahydroxy-5-oxohexanoic acid
SMILES OCC(=O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H10O7/c7-1-2(8)3(9)4(10)5(11)6(12)13/h3-5,7,9-11H,1H2,(H,12,13)/t3-,4+,5-/m1/s1
Formula Weight 194.0427
Chemical Name or Material 5-Keto-D-gluconic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.