CAS RN 5287-64-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Keto-D-gluconic Acid, TRC
CAS: 5287-64-9 Molecular Formula: C6H10O7 Molecular Weight (g/mol): 194.14 Synonym: D-xylo-5-Hexulosonic acid,5-Oxo-D-gluconic acid,5-Keto-D-gluconic acid,5-keto-D-Gluconic acid IUPAC Name: (2R,3S,4S)-2,3,4,6-tetrahydroxy-5-oxohexanoic acid SMILES: OCC(=O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O