missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-3-methyl-2-methylaminopyridine, 95%, Thermo Scientific™

Product Code. p-7030171
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15403767 250 mg 250mg
11372839 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15403767 Supplier Thermo Scientific Alfa Aesar Supplier No. H27718.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 942206-33-9
Molecular Formula C7H9IN2
Molecular Weight (g/mol) 248.07
MDL Number MFCD09037456
InChI Key ZHUKYUJCPFBDHP-UHFFFAOYSA-N
Synonym 5-iodo-3-methyl-2-methylaminopyridine, 5-iodo-n,3-dimethyl-2-pyridinamine
PubChem CID 37030063
IUPAC Name 5-iodo-N,3-dimethylpyridin-2-amine
SMILES CNC1=NC=C(I)C=C1C

Specifications

Melting Point 70°C to 73°C
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 248.07
Percent Purity 95%
Chemical Name or Material 5-Iodo-3-methyl-2-methylaminopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.