accessibility menu, dialog, popup

UserName

CH3NHNCH3I

CAS RN 942206-33-9

IUPAC Name: 5-iodo-N,3-dimethylpyridin-2-amine
Synonyms: 5-iodo-N,3-dimethylpyridin-2-amine
Molecular Weight (g/mol): 248.07
Molecular Formula: C7H9IN2
InChi Key: ZHUKYUJCPFBDHP-UHFFFAOYSA-N
SMILES: CNC1=NC=C(I)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results