missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyhexanoic Acid Sodium Salt, TRC

Product Code. 30390602
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390602 250 mg -
30391919 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30390602 Supplier Toronto Research Chemicals Supplier No. TRCH942825250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 185956-02-9
Molecular Formula C6H11NaO3
Molecular Weight (g/mol) 154.14
Synonym 5-Hydroxycaproic Acid
IUPAC Name sodium 5-hydroxyhexanoate
SMILES OC(CCCC(O[Na])=O)C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C6H12O3.Na/c1-5(7)3-2-4-6(8)9;/h5,7H,2-4H2,1H3,(H,8,9);/q;+1/p-1
Formula Weight 154.06
Chemical Name or Material 5-Hydroxyhexanoic Acid Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.