CAS RN 185956-02-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Hydroxyhexanoic Acid Sodium Salt, TRC
CAS: 185956-02-9 Molecular Formula: C6H11NaO3 Molecular Weight (g/mol): 154.14 Synonym: 5-Hydroxycaproic Acid IUPAC Name: sodium 5-hydroxyhexanoate SMILES: OC(CCCC(O[Na])=O)C