missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy Thiabendazole Methyl Ether Hydrochloride, TRC

Product Code. 30403643
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403643 2.5 mg 2.5mg
30396764 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30403643 Supplier Toronto Research Chemicals Supplier No. TRCH9612052.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2318-79-8
Molecular Formula C11H9N3OS . HCl
Molecular Weight (g/mol) 267.74
Synonym 5-Methoxy-2-(4-thiazolyl)-benzimidazole Monohydrochloride
IUPAC Name 4-(5-methoxy-1H-benzimidazol-2-yl)-1,3-thiazole;hydrochloride
SMILES Cl.COc1ccc2[nH]c(nc2c1)c3cscn3

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C11H9N3OS.ClH/c1-15-7-2-3-8-9(4-7)14-11(13-8)10-5-16-6-12-10;/h2-6H,1H3,(H,13,14);1H
Formula Weight 267.0233
Chemical Name or Material 5-Hydroxy Thiabendazole Methyl Ether Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.