CAS RN 2318-79-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Hydroxy Thiabendazole Methyl Ether Hydrochloride, TRC
CAS: 2318-79-8 Molecular Formula: C11H9N3OS . HCl Molecular Weight (g/mol): 267.74 Synonym: 5-Methoxy-2-(4-thiazolyl)-benzimidazole Monohydrochloride IUPAC Name: 4-(5-methoxy-1H-benzimidazol-2-yl)-1,3-thiazole;hydrochloride SMILES: Cl.COc1ccc2[nH]c(nc2c1)c3cscn3