missing translation for 'onlineSavingsMsg'
Learn More

5-Hex-1-ynylnicotinic acid, 97%, Thermo Scientific™

Product Code. 10408914
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10408914 5 g 5g
10161312 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10408914 Supplier Thermo Scientific Maybridge Supplier No. KM09434DE
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 306935-31-9
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD00830967
InChI Key FKWDNFDBDZUPIQ-UHFFFAOYSA-N
Synonym 5-hex-1-ynylnicotinic acid, 5-hex-1-ynylnicotinc acid, 3-pyridinecarboxylicacid, 5-1-hexyn-1-yl, 5-hex-1-yn-1-yl pyridine-3-carboxylic acid, 5-hex-1-ynyl-nicotinic acid, 5-hex-1-ynyl nicotinic acid, 5-hex-1-yn-1-yl nicotinic acid
PubChem CID 2736516
IUPAC Name 5-hex-1-ynylpyridine-3-carboxylic acid
SMILES CCCCC#CC1=CC(=CN=C1)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 203.24
Percent Purity 97%
Chemical Name or Material 5-Hex-1-ynylnicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.