accessibility menu, dialog, popup

UserName

H3CNOOH

CAS RN 306935-31-9

IUPAC Name: 5-(hex-1-yn-1-yl)pyridine-3-carboxylic acid
Synonyms: 5-(hex-1-yn-1-yl)pyridine-3-carboxylic acid
Molecular Weight (g/mol): 203.24
Molecular Formula: C12H13NO2
InChi Key: FKWDNFDBDZUPIQ-UHFFFAOYSA-N
SMILES: CCCCC#CC1=CC(=CN=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results