missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroindole-3-butyric Acid, TRC

Product Code. 30393904
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393904 100 mg
30401526 250 mg
30401952 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393904 Supplier Toronto Research Chemicals Supplier No. TRCF402818100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 319-72-2
Molecular Formula C12H12FNO2
Molecular Weight (g/mol) 221.23
Synonym 4-(5-Fluoro-1H-indol-3-yl)butanoic Acid
SMILES OC(=O)CCCc1c[nH]c2ccc(F)cc12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H12FNO2/c13-9-4-5-11-10(6-9)8(7-14-11)2-1-3-12(15)16/h4-7,14H,1-3H2,(H,15,16)
Formula Weight 221.0852
Chemical Name or Material 5-Fluoroindole-3-butyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.