CAS RN 319-72-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Fluoroindole-3-butyric Acid, TRC
CAS: 319-72-2 Molecular Formula: C12H12FNO2 Molecular Weight (g/mol): 221.23 Synonym: 4-(5-Fluoro-1H-indol-3-yl)butanoic Acid SMILES: OC(=O)CCCc1c[nH]c2ccc(F)cc12