missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-(trifluoromethoxy)benzoyl chloride, 97%, Thermo Scientific™

Product Code. 15475567
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475567 1 g 1g
15438205 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475567 Supplier Thermo Scientific Alfa Aesar Supplier No. H32804.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1092460-84-8
Molecular Formula C8H3ClF4O2
Molecular Weight (g/mol) 242.554
MDL Number MFCD11519334
InChI Key ZZRULKQBGZXHPC-UHFFFAOYSA-N
Synonym 5-fluoro-2-trifluoromethoxy benzoyl chloride, 5-fluoro-2-trifluoromethoxy benzoyl chloride, jrd
PubChem CID 57364110
IUPAC Name 5-fluoro-2-(trifluoromethoxy)benzoyl chloride
SMILES C1=CC(=C(C=C1F)C(=O)Cl)OC(F)(F)F

Specifications

Refractive Index 1.4559
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 242.55
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-(trifluoromethoxy)benzoyl chloride
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.