accessibility menu, dialog, popup

UserName

CAS RN 1092460-84-8

FOClOFFF

CAS RN 1092460-84-8

IUPAC Name: 5-fluoro-2-(trifluoromethoxy)benzoyl chloride
Synonyms: 5-fluoro-2-(trifluoromethoxy)benzoyl chloride
Molecular Weight (g/mol): 242.55
Molecular Formula: C8H3ClF4O2
InChi Key: ZZRULKQBGZXHPC-UHFFFAOYSA-N
SMILES: FC1=CC(C(Cl)=O)=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results