missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylbenzothiazole, TRC

Product Code. 30400152
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400152 250 mg
30397344 100 mg
30406596 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400152 Supplier Toronto Research Chemicals Supplier No. TRCF597040250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 399-75-7
Molecular Formula C8H6FNS
Molecular Weight (g/mol) 167.2
IUPAC Name 5-fluoro-2-methyl-1,3-benzothiazole
SMILES Cc1nc2cc(F)ccc2s1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C8H6FNS/c1-5-10-7-4-6(9)2-3-8(7)11-5/h2-4H,1H3
Formula Weight 167.0205
Chemical Name or Material 5-Fluoro-2-methylbenzothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.