CAS RN 399-75-7
IUPAC Name:
5-fluoro-2-methyl-1,3-benzothiazole
Synonyms:
5-fluoro-2-methyl-1,3-benzothiazole
Molecular Weight (g/mol):
167.2
Molecular Formula:
C8H6FNS
InChi Key:
LDBFGRGBYVULTJ-UHFFFAOYSA-N
SMILES:
CC1=NC2=CC(F)=CC=C2S1
1
–
1
of
1
results
1
5-Fluoro-2-methylbenzothiazole, TRC
CAS: 399-75-7 Molecular Formula: C8H6FNS Molecular Weight (g/mol): 167.2 IUPAC Name: 5-fluoro-2-methyl-1,3-benzothiazole SMILES: Cc1nc2cc(F)ccc2s1