missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methoxybenzoic acid, 97%, Thermo Scientific™

Product Code. 15485107
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485107 5 g 5g
15475107 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15485107 Supplier Thermo Scientific Alfa Aesar Supplier No. H32389.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Fluoro-3-hydroxy-2-methylbenzoic acid 1 was synthesized from 5-fluoro-2-methylbenzoic acid by nitration, reduction, diazotization and hydrolysis. Another methylcarbamoyl-substituted benzylamine 12 was synthesized from 5-fluoro-2-methylbenzoic acid via the sequential bromination,azido replacement and the reduction.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 394-04-7
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD00671765
InChI Key WPXFJBPJUGMYOD-UHFFFAOYSA-N
Synonym 5-fluoro-2-methoxy-benzoic acid, 2-methoxy-5-fluorobenzoic acid, benzoic acid, 5-fluoro-2-methoxy, 5-fluoro-2-methoxylbenzoic acid, pubchem2677, 5-fluoro-o-anisic acid, 2-carboxy-4-fluoroanisole, rarechem al bo 1468, buttpark 32\01-79
PubChem CID 2737364
IUPAC Name 5-fluoro-2-methoxybenzoic acid
SMILES COC1=C(C=C(C=C1)F)C(=O)O

Specifications

Melting Point 86°C to 90°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 170.14
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-methoxybenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.