accessibility menu, dialog, popup

UserName

H3COFOOH

CAS RN 394-04-7

IUPAC Name: 5-fluoro-2-methoxybenzoic acid
Synonyms: 5-fluoro-2-methoxybenzoic acid
Molecular Weight (g/mol): 170.14
Molecular Formula: C8H7FO3
InChi Key: WPXFJBPJUGMYOD-UHFFFAOYSA-N
SMILES: COC1=C(C=C(F)C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results