missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methoxybenzaldehyde, 98%

Product Code. 11460558
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460558 25 g 25g
11440558 1 g 1g
11450558 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460558 Supplier Thermo Scientific Alfa Aesar Supplier No. B24684.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19415-51-1 | C8H7FO2 | 154.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19415-51-1
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00143458
InChI Key CRLDWFVRQNUUSZ-UHFFFAOYSA-N
PubChem CID 2734943
IUPAC Name 5-fluoro-2-methoxybenzaldehyde
SMILES COC1=C(C=C(C=C1)F)C=O

Specifications

Quantity 25 g
Sensitivity Air Sensitive
Synonym 2-methoxy-5-fluorobenzaldehyde, 5-fluoro-o-anisaldehyde, 5-fluoro-o-methoxybenzaldehyde, benzaldehyde, 5-fluoro-2-methoxy, 5-fluoro-2-methoxy-benzaldehyde, 5-fluoro-o-2-methoxybenzaldehyde, pubchem2636, acmc-209exz, 2-methoxy-5-fluoro benzaldehyde, benzaldehyde,5-fluoro-2-methoxy
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 5-Fluoro-2-methoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.