CAS RN 19415-51-1
IUPAC Name:
5-fluoro-2-methoxybenzaldehyde
Synonyms:
5-fluoro-2-methoxybenzaldehyde
Molecular Weight (g/mol):
154.14
Molecular Formula:
C8H7FO2
InChi Key:
CRLDWFVRQNUUSZ-UHFFFAOYSA-N
SMILES:
COC1=CC=C(F)C=C1C=O
1
–
1
of
1
results
1
5-Fluoro-2-methoxybenzaldehyde, 98%
CAS: 19415-51-1 Molecular Formula: C8H7FO2 Molecular Weight (g/mol): 154.14 MDL Number: MFCD00143458 InChI Key: CRLDWFVRQNUUSZ-UHFFFAOYSA-N PubChem CID: 2734943 IUPAC Name: 5-fluoro-2-methoxybenzaldehyde SMILES: COC1=C(C=C(C=C1)F)C=O