missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-hydroxy-3-nitropyridine, 98%, Thermo Scientific™

Product Code. 15491649
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491649 5 g 5g
15401659 250 mg 250mg
15481649 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491649 Supplier Thermo Scientific Alfa Aesar Supplier No. H64441.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136888-20-5
Molecular Formula C5H3FN2O3
Molecular Weight (g/mol) 158.088
MDL Number MFCD05662412
InChI Key BLFUHTOZIOQGBU-UHFFFAOYSA-N
Synonym 5-fluoro-2-hydroxy-3-nitropyridine, 5-fluoro-3-nitropyridin-2-ol, 5-fluoro-3-nitropyridin-2 1h-one, 2-hydroxy-3-nitro-5-fluoropyridine, 2-hydroxy-5-fluoro-3-nitropyridine, 2 1h-pyridinone, 5-fluoro-3-nitro, pubchem2975, acmc-1cgzr, ksc520k3n, 5-fluoro-2-hydroxy-3-nitrop
PubChem CID 10583047
IUPAC Name 5-fluoro-3-nitro-1H-pyridin-2-one
SMILES C1=C(C(=O)NC=C1F)[N+](=O)[O-]

Specifications

Quantity 5 g
Beilstein 7582013
Solubility Information Slightly soluble in water.
Formula Weight 158.09
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Fluoro-2-hydroxy-3-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.