accessibility menu, dialog, popup

UserName

OHNFNOO

CAS RN 136888-20-5

IUPAC Name: 5-fluoro-3-nitro-1,2-dihydropyridin-2-one
Synonyms: 5-fluoro-3-nitro-1H-pyridin-2-one
Molecular Weight (g/mol): 158.09
Molecular Formula: C5H3FN2O3
InChi Key: BLFUHTOZIOQGBU-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=CNC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results