missing translation for 'onlineSavingsMsg'
Learn More

5-Ethyl-demethyl Lercanidipine, TRC

Product Code. 30385790
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385790 100 mg
30369669 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385790 Supplier Toronto Research Chemicals Supplier No. TRCE913200100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 786625-22-7
Molecular Formula C37H43N3O6
Molecular Weight (g/mol) 625.75
Synonym 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 3-[2-[(3,3-Diphenylpropyl)methylamino]-1,1-dimethylethyl] 5-ethyl Ester, 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 2-[(3,3-Diphenylpropyl)methylamino]-1,1-dimethylethyl Ethyl Ester
SMILES CCOC(=O)C1=C(C)NC(=C(C1c2cccc(c2)[N+](=O)[O-])C(=O)OC(C)(C)CN(C)CCC(c3ccccc3)c4ccccc4)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C37H43N3O6/c1-7-45-35(41)32-25(2)38-26(3)33(34(32)29-19-14-20-30(23-29)40(43)44)36(42)46-37(4,5)24-39(6)22-21-31(27-15-10-8-11-16-27)28-17-12-9-13-18-28/h8-20,23,31,34,38H,7,21-22,24H2,1-6H3
Formula Weight 625.3152
Chemical Name or Material 5-Ethyl-demethyl Lercanidipine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.