CAS RN 786625-22-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Ethyl-demethyl Lercanidipine, TRC
CAS: 786625-22-7 Molecular Formula: C37H43N3O6 Molecular Weight (g/mol): 625.75 Synonym: 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 3-[2-[(3,3-Diphenylpropyl)methylamino]-1,1-dimethylethyl] 5-ethyl Ester,1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 2-[(3,3-Diphenylpropyl)methylamino]-1,1-dimethylethyl Ethyl Ester SMILES: CCOC(=O)C1=C(C)NC(=C(C1c2cccc(c2)[N+](=O)[O-])C(=O)OC(C)(C)CN(C)CCC(c3ccccc3)c4ccccc4)C