missing translation for 'onlineSavingsMsg'
Learn More

5-(Diphenylphosphinyl)pentanoic Acid, TRC

Product Code. 30385296
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385296 250 mg 250mg
30371528 50 mg -
30383822 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385296 Supplier Toronto Research Chemicals Supplier No. TRCD492135250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71140-70-0
Molecular Formula C17H19O3P
Molecular Weight (g/mol) 302.3
SMILES OC(=O)CCCCP(=O)(c1ccccc1)c2ccccc2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H19O3P/c18-17(19)13-7-8-14-21(20,15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-6,9-12H,7-8,13-14H2,(H,18,19)
Formula Weight 302.1072
Chemical Name or Material 5-(Diphenylphosphinyl)pentanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.