CAS RN 71140-70-0
CAS RN 71140-70-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-(Diphenylphosphinyl)pentanoic Acid, TRC
CAS: 71140-70-0 Molecular Formula: C17H19O3P Molecular Weight (g/mol): 302.3 SMILES: OC(=O)CCCCP(=O)(c1ccccc1)c2ccccc2