missing translation for 'onlineSavingsMsg'
Learn More

5-(Dihydroxyboryl)-2-thiophenecarboxylic acid, 95+%, Maybridge™

Product Code. 10255364
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10255364 10 g 10g
10138264 5 g 5g
10487544 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10255364 Supplier Thermo Scientific Maybridge Supplier No. MO00596EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 465515-31-5
Molecular Formula C5H5BO4S
Molecular Weight (g/mol) 171.961
InChI Key OQGIKNPOYTVNNF-UHFFFAOYSA-N
Synonym 2-carboxythiophene-5-boronic acid, 5-carboxythiophene-2-boronic acid, 5-dihydroxyboryl-2-thiophenecarboxylic acid, 5-dihydroxyboryl thiophene-2-carboxylic acid, 5-dihydroxyboranyl thiophene-2-carboxylic acid, 5-borono-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-borono, pubchem7851, acmc-1ap1p, d0jv5j
PubChem CID 2779807
IUPAC Name 5-boronothiophene-2-carboxylic acid
SMILES B(C1=CC=C(S1)C(=O)O)(O)O

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 171.97
Percent Purity 95%
Chemical Name or Material 5-(Dihydroxyboryl)-2-thiophenecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.