CAS RN 465515-31-5
IUPAC Name:
5-(dihydroxyboranyl)thiophene-2-carboxylic acid
Synonyms:
5-(dihydroxyboranyl)thiophene-2-carboxylic acid
Molecular Weight (g/mol):
171.96
Molecular Formula:
C5H5BO4S
InChi Key:
OQGIKNPOYTVNNF-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC=C(S1)C(O)=O
1
–
1
of
1
results
1
5-(Dihydroxyboryl)-2-thiophenecarboxylic acid, 95+%, Maybridge™
CAS: 465515-31-5 Molecular Formula: C5H5BO4S Molecular Weight (g/mol): 171.961 InChI Key: OQGIKNPOYTVNNF-UHFFFAOYSA-N Synonym: 2-carboxythiophene-5-boronic acid,5-carboxythiophene-2-boronic acid,5-dihydroxyboryl-2-thiophenecarboxylic acid,5-dihydroxyboryl thiophene-2-carboxylic acid,5-dihydroxyboranyl thiophene-2-carboxylic acid,5-borono-2-thiophenecarboxylic acid,2-thiophenecarboxylic acid, 5-borono,pubchem7851,acmc-1ap1p,d0jv5j PubChem CID: 2779807 IUPAC Name: 5-boronothiophene-2-carboxylic acid SMILES: B(C1=CC=C(S1)C(=O)O)(O)O