missing translation for 'onlineSavingsMsg'
Learn More

5-Cyanopentylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15475365
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475365 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15475365 Supplier Thermo Scientific Alfa Aesar Supplier No. H58480.AD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Reacts with water.

Notes
Air Sensitive. Incompatible with oxidizing agents, acid chlorides. Store under dry inert gas. Protect from humidity and moisture. Store away from air.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1443250-11-0
Molecular Formula C6H10BrNZn
Linear Formula N≡C(CH2)5ZnBr
MDL Number MFCD08741503
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym 5-cyanovalerylzinc bromide 0.5 m in tetrahydrofuran, 5-cyanovalerylzinc bromide, 0.5m in tetrahydrofuran, 5-cyanopentylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
Solubility Information Reacts with water.
InChI Key UODGVECCRVRLMB-UHFFFAOYSA-M
SMILES [CH2-]CCCCC#N.[Zn+]Br
IUPAC Name bromozinc(1+);hexanenitrile
Molecular Weight (g/mol) 241.437
PubChem CID 11425010
Formula Weight 241.44
Chemical Name or Material 5-Cyanopentylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.