missing translation for 'onlineSavingsMsg'
Learn More

5-Chloropentyl acetate, 90+%

Product Code. 11458993
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458993 50 g 50g
11448993 10 g 10g
11468993 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458993 Supplier Thermo Scientific Alfa Aesar Supplier No. A11111.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 20395-28-2 | C7H13ClO2 | 164.629 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20395-28-2
Molecular Formula C7H13ClO2
Molecular Weight (g/mol) 164.629
MDL Number MFCD00013696
InChI Key ZCYVIAZIVJNAMO-UHFFFAOYSA-N
Synonym 5-chloro-n-amyl acetate, 1-acetoxy-5-chloropentane, 5-chloro-1-pentanol acetate, 5-chloro-1-amyl acetate, 5-chloro-1-acetoxypentane, 1-pentanol, 5-chloro-, acetate, 5-chloro-1-pentyl acetate, 1-pentanol, 5-chloro-, 1-acetate, 5-chloroamyl acetate, acetic acid 5-chloro-pentyl ester
PubChem CID 88521
IUPAC Name 5-chloropentyl acetate
SMILES CC(=O)OCCCCCCl

Specifications

Density 1.061
Boiling Point 96°C to 98°C (11 mmHg)
Flash Point 97°C (206°F)
Refractive Index 1.438
Quantity 50 g
Beilstein 1752107
Formula Weight 164.63
Percent Purity 98%
Chemical Name or Material 5-Chloropentyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.